Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRG-L2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA146144

Catalog No. PIPA146144


Add to cart

Description

Description

The predicted MW is 48 kDa; however the protein runs above 50 kDa on gels and can form dimers.

A novel gene, CRG-L2 (Cancer related gene-Liver 2) displays high expression in murine and human hepatocellular carcinomas (HCC). Its restricted expression in normal tissues and unique early upregulation during tumor development make CRG-L2 an excellent candidate as a new clinical marker of HCC.
Specifications

Specifications

CRG-L2
Polyclonal
Unconjugated
GLDN
Liquid
GLDN
Residues 160-176 of human CRG-L2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q6ZMI3
CLOM, COLM, CRG-L2, CRGL2, FLJ23917, UNC-112
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
342035
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit