Learn More
CRIP1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA524643
Catalog No. PIPA524643
Add to cart
Description
Description
This antibody is predicted to react with bovine and rat based on sequence homology.
Cysteine-rich intestinal protein (CRIP) belongs to the LIM/double zinc finger protein family, members of which include cysteine- and glycine-rich protein-1 (CSRP1; MIM 123876), rhombotin-1 (RBTN1; MIM 186921), rhombotin-2 (RBTN2; MIM 180385), and rhombotin-3 (RBTN3; MIM 180386). CRIP may be involved in intestinal zinc transport.
Specifications
Specifications
CRIP1
Polyclonal
Unconjugated
CRIP1
Liquid
CRIP1
KLH conjugated synthetic peptide between 48-77 amino acids from the C-terminal region of human CRIP1
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human, Mouse
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
P50238, P63254
CRHP, CRIP, CRP-1, CRP1, cysteine-rich heart protein, cysteine-rich intestinal protein, cysteine-rich protein 1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
12925, 1396