Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRISP-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP179533
Description
CRISP-1 Polyclonal specifically detects CRISP-1 in Human samples. It is validated for Western Blot.Specifications
CRISP-1 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
Acidic epididymal glycoprotein homolog, acidic epididymal glycoprotein-like 1, AEG-related protein, ARPAEG-like protein, CRISP-1AEGL1HSCRISP1D, cysteine-rich secretory protein 1, cysteine-rich secretory protein-1 delta, HSCRISP1G, HUMARP | |
Rabbit | |
Affinity purified | |
RUO | |
167 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
NP_001122 | |
CRISP1 | |
Synthetic peptide directed towards the N terminal of human CRISP1The immunogen for this antibody is CRISP1. Peptide sequence LKMSWSEEAAQNARIFSKYCDMTESNPLERRLPNTFCGENMHMTSYPVSW. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Bovine: 84%; Goat: 84%; Pig: 84%; Rabbit: 78%. | |
Human, Porcine, Bovine, Canine, Goat, Rabbit, Zebrafish | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction