Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRISPLD2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525482

Catalog No. PIPA525482

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

CRISPLD2 promotes matrix assembly.
Specifications

Specifications

CRISPLD2
Polyclonal
PBS with 0.09% sodium azide
Q9H0B8
CRISP11, LCRISP2, CRISP-11, LCCL domain containing cysteine-rich secretory protein 2, LCCL domain-containing cysteine-rich secretory protein 2, cysteine-rich secretory protein 11, cysteine-rich secretory protein LCCL domain-containing 2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
83716
Western Blot
Unconjugated
CRISPLD2
Liquid
CRISPLD2
KLH conjugated synthetic peptide between 31-58 amino acids from the N-terminal region of human CRISPLD2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit