Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRM1 Rabbit anti-Bovine, Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA1083

Catalog No. PA1083


Add to cart

Description

Description

PA1-083 detects CRM1 (chromosome region maintece 1) from bovine retinal extract, human and mouse samples. PA1-083 has been successfully used in Western blot and immunoprecipitation procedures. By Western blot, this antibody detects an ~135 kDa protein representing CRM1 from bovine retinal extract. The PA1-083 immunogen is a synthetic peptide corresponding to residues C R(1043) Q A D E E K H K R Q M S V P G S(1058) of human CRM1. This sequence is 88% conserved in mouse, rat, and Xenopus laevis. The PA1-083 immunizing peptide (Cat. No. PEP-162) is available for use in neutralization and control experiments.

Diffusion of metabolites and small non-nuclear molecules as well as active, mediated import of protein and export of protein and RNA through the nuclear envelope occurs through nuclear pore complexes or NPC’s. NPC’s contain up to 100 different polypeptides which have a combined mass of about 125 megadaltons. The channel available for passive transport through the NPC is about 9-10 nm in diameter while carrier mediated changes in the NPC result in a ~25 nm channel used for larger, actively transported molecules.   Active transport of molecules through the NPC is a signal-mediated process that is not well understood. Proteins to be exported from the nucleus to cytoplasm contain leucine-rich nuclear export signals (NESs). NESs are recognized by transport receptors belonging to the importin-beta family of proteins. It has been shown that NES export is mediated by CRM1 (chromosome region maintece 1), an importin-beta family member that binds NESs, Ran:GTP, and Ran binding protein (RanBP). CRM1 mediates the nuclear exportation of numerous proteins, including histone deacetylases (HDACs), viral proteins, immunophilins, and STAT1.

Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly

Specifications

Specifications

CRM1
Polyclonal
Unconjugated
XPO1
Liquid
XPO1
Synthetic peptide corresponding to residues C R(1043) Q A D E E K H K R Q M S V P G S(1058) of human CRM1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Bovine, Human, Mouse
Immunoprecipitation, Western Blot
1 mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide
O14980, Q6P5F9
Chromosome Region Maintece 1
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
103573, 510568, 7514
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit