Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRMP1 Mouse anti-Human, Clone: Y21, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 05706
Description
This Anti-CRMP1 Antibody, clone Y21 is validated for use in western blotting for the detection of CRMP1.Specifications
CRMP1 | |
Monoclonal | |
Unconjugated | |
NM_001313 | |
Mouse | |
Ascites | |
Primary | |
−20°C for 24 months. | |
IgM κ |
Western Blot | |
Y21 | |
mouse ascites with 0.05% sodium azide before the addition of glycerol to 30% | |
CRMP1, DPYSL1, DRP1, CRMP-1, DRP-1 | |
His6-tagged fusion protein corresponding to amino acids 76-572 of human CRMP1 | |
100 μL | |
Human | |
Ascites |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction