Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CROT Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526386

Catalog No. PIPA526386


Add to cart

Description

Description

CROT encodes a member of the carnitine/choline acetyltransferase family. The encoded protein converts 4,8-dimethylnooyl-CoA to its corresponding carnitine ester. This transesterification occurs in the peroxisome and is necessary for transport of medium- and long- chain acyl-CoA molecules out of the peroxisome to the cytosol and mitochondria. The protein thus plays a role in lipid metabolism and fatty acid beta-oxidation. Alternatively spliced transcript variants have been described.
Specifications

Specifications

CROT
Polyclonal
PBS with 0.09% sodium azide
Q9UKG9, Q9DC50
COT, peroxisomal carnitine O-octanoyltransferase, peroxisomal carnitine acyltransferase
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
54677, 74114
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
CROT
Liquid
CROT
KLH conjugated synthetic peptide between 49-79 amino acids from the N-terminal region of human CROT
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit