Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRTAC1 Mouse anti-Human, Clone: 755315, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA524362

Catalog No. PIMA524362


Add to cart

Description

Description

Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.

Specifications

Specifications

CRTAC1
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
Q9NQ79
ASPIC1, CEP68, ASPIC, ASPIC1CEP-68acidic secreted protein in cartilage, cartilage acidic protein 1, CEP68, chondrocyte expressed protein 68 kDa CEP-68,68 kDa chondrocyte-expressed protein, FLJ10320
Mouse
IgG2B
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
55118
Immunocytochemistry, Immunofluorescence
755315
Unconjugated
CRTAC1
Lyophilized
CRTAC1
Mouse myeloma cell line NS0-derived recombit human CRTAC1 Ser28-Cys661
Protein A/G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit