Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

crystallin, beta A2, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human CRYBA2 protein.

Manufacturer:  Abnova Corporation H00001412B01

Catalog No. 89-005-177


Add to cart

Description

Description

Crystallins are separated into two classes: taxon-specific, or enzyme, and ubiquitous. The latter class constitutes the major proteins of the vertebrate eye, which function to maintain the transparency and refractive index of the lens. Since lens central fiber cells lose their nuclei during development, these crystallins are made and then retained throughout life, making them extremely stable proteins. Mammalian lens crystallins are divided into alpha, beta, and gamma families; beta and gamma crystallins are also defined as a superfamily. Alpha and beta families are further divided into acidic and basic groups. Seven protein regions exist in crystallins: four homologous motifs, a connecting peptide, and N- and C-terminal extensions. Beta-crystallins, the most heterogeneous, differ by the presence of the C-terminal extension (present in the basic group but absent in the acidic group). Beta-crystallins form aggregates of different sizes and are able to form homodimers through self-association or heterodimers with other beta-crystallins. This gene is a beta acidic group member. Three alternatively spliced transcript variants encoding identical proteins have been reported. [provided by RefSeq

Sequence: MSSAPAPGPAPASLTLWDEEDFQGRRCRLLSDCANVCERGGLPRVRSVKVENGVWVAFEYPDFQGQQFILEKGDYPRWSAWSGSSSHNSNQLLSFRPVLCANHNDSRVTLFEGDNFQGCKFDLVDDYPSLPSMGWASKDVGSLKVSSGAWVAYQYPGYRGYQYVLERDRHSGEFCTYGELGTQAHTGQLQSIRRVQH
Specifications

Specifications

crystallin, beta A2
Unconjugated
No additive
BC006285
CRYBA2
CRYBA2 (AAH06285, 1 a.a. ~ 197 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
Western Blot
Mouse polyclonal antibody raised against a full-length human CRYBA2 protein.
CRYBA2
Serum
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
1412
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit