Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NLRP3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA521745

Catalog No. PIPA521745


Add to cart

Description

Description

PA5-21745 targets Cryopyrin in WB applications and shows reactivity with Human samples. The PA5-21745 immunogen is a synthetic peptide corresponding to a region within amino acids 974 and 1036 of NLRP3 (Uniprot IDNo.Q96P20). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

NLRP3
Polyclonal
Unconjugated
NLRP3
Liquid
NLRP3
Synthetic peptide corresponding to a region within amino acids 974 and 1036 of NLRP3 (Uniprot IDNo.Q96P20)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q96P20
AGTAVPRL, AII, AVP, C1orf7, CIAS1, CLR1.1, FCAS, FCU, MWS, NALP3, PYPAF1, NACHT domain-, leucine-rich repeat-, and PYD-containing protein 3, NACHT, LRR and PYD containing protein 3, NACHT, LRR and PYD domains-containing protein 3, PYRIN-containing APAF1-like protein 1, caterpiller protein 1.1, cold autoinflammatory syndrome 1 protein, cryopyrin, nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 3
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
114548
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit