Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cryptosporidium parvum Oocysts Goat anti-Protozoa, HRP, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA173185

Catalog No. PA173185


Add to cart

Description

Description

The PA1-73185 antibody reacts with Cryptosporidium parvum oocysts. Crossreacts with a large rod-shaped microorganism found in bovine feces. PA1-73185 has been successfully used in ELISA applications. The PA1-73185 antibody was raised against oocysts purified from bovine feces. NOTE: sodium azide is a potent inhibitor of peroxidase!

Specifications

Specifications

Cryptosporidium parvum Oocysts
Polyclonal
HRP
Liquid
Goat
IgG
1 mL
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Primary
Protozoa
ELISA
1-2 mg/mL
PBS with 10mg/mL BSA and 0.002% thimerosal
Cryptosporidium parvum; Cryptosporidiosis
Oocysts purified from bovine feces.
Purified
RUO
Antibody
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit