Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cryptosporidium parvum Oocysts Goat anti-Protozoa, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA173183

Catalog No. PA173183


Add to cart

Description

Description

The PA1-73183 antibody reacts with Cryptosporidium parvum oocysts. PA1-73183 has been successfully used in ELISA and Immunofluorescencel applications. The PA1-73183 antibody was raised against oocysts purified from bovine feces.

Specifications

Specifications

Cryptosporidium parvum Oocysts
Polyclonal
Unconjugated
Liquid
Goat
IgG
1 mL
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Primary
Protozoa
ELISA, Immunofluorescence
4-5 mg/mL
PBS with 0.1% sodium azide; pH 7.2
Cryptosporidium parvum; Cryptosporidiosis
Oocysts purified from bovine feces.
Purified
RUO
Antibody
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit