Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAG3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525339

Catalog No. PIPA525339


Add to cart

Description

Description

The expression of the cancer-testis antigen Taxol resistanceassociated gene-3 (TRAG-3) is associated with acquired paclitaxel (Taxol) resistance, and is expressed in a variety of cancers - e.g., breast cancer, leukemia, and melanoma. Overexpressed in taxolresistant breast cancer line MDA 435(TR) and the doxorubicinresistant multiple myelanoma lines 8226/Dox(40) and 8226/MDR (10)V. Weakly expressed in kidney. TRAG-3 is an attractive target for immunotherapy of cancer.
Specifications

Specifications

TRAG3
Polyclonal
PBS with 0.09% sodium azide
Q9Y5P2
CSAG3, CSAG3B, CT24.2, TRAG-3, TRAG3, CSAG family, member 3B, cancer/testis antigen 24.2, cancer/testis antigen family 24, member 2, chondrosarcoma-associated gene 2/3 protein, taxol resistance associated gene 3, taxol-resistant-associated gene 3 protein
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
102723547
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
CSAG2
Liquid
CSAG2
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human CSAG2
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit