Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CAS Monoclonal Antibody (P729), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA518303
Description
Recommended positive controls: 293T, A431, HeLa, HepG2, Neuro2A, GL261, C8D30, NIH-3T3, BCL-1, Raw264.7, C2C12, PC-12, Rat2. Predicted reactivity: Mouse (99%), Rat (100%), Zebrafish (95%), Xenopus laevis (97%), Pig (100%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CAS Monoclonal antibody specifically detects CAS in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blot.Specifications
CAS | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
P55060, Q9ERK4 | |
CSE1L | |
Recombinant fragment corresponding to a region within amino acids 222 and 469 of CSE1L | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG2a |
Immunocytochemistry, Western Blot | |
P729 | |
Unconjugated | |
CSE1L | |
2610100P18Rik; AA407533; Capts; CAS; Cellular apoptosis susceptibility protein; chromosome segregation 1 like; chromosome segregation 1-like (S. cerevisiae); chromosome segregation 1-like protein; CSE1; CSE1 chromosome segregation 1 like; CSE1 chromosome segregation 1-like; CSE1 chromosome segregation 1-like (yeast); CSE1L; Exp2; exportin-2; Importin-alpha re-exporter; MGC117283; MGC130036; MGC130037; XPO2 | |
Mouse | |
Protein G | |
RUO | |
110750, 1434, 362273 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction