Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAS Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA521468

Catalog No. PIPA521468


Add to cart

Description

Description

PA5-21468 targets CSE1L in immunocytochemistry, immunofluorescence, immunohistochemistry (paraffin), and Western blot applications and shows reactivity with Human, mouse, and rat samples. The PA5-21468 immunogen is a recombit fragment corresponding to a region within amino acids 222 and 469 of CSE1L (Uniprot IDNo.P55060). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

CAS
Polyclonal
Unconjugated
CSE1L
Liquid
CSE1L
Recombit fragment corresponding to a region within amino acids 222 and 469 of CSE1L (Uniprot IDNo.P55060)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.59 mg/mL
PBS with 20% glycerol and 0.025% Proclin 300; pH 7
P55060, Q9ERK4
CAS, CSE1, XPO2, cellular apoptosis susceptibility protein, chromosome segregation 1-like protein, exp2, exportin-2, importin-alpha re-exporter
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
110750, 1434, 362273
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit