Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KChIP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA528330

Catalog No. PIPA528330


Add to cart

Description

Description

PA5-28330 targets CSEN in WB applications and shows reactivity with Human and mouse samples. The PA5-28330 immunogen is recombit fragment corresponding to a region within amino acids 1 and 256 of Human CSEN. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

This gene encodes a member of the family of voltage-gated potassium (Kv) channel-interacting proteins, which belong to the recoverin branch of the EF-hand superfamily. Members of this family are small calcium binding proteins containing EF-hand-like domains. They are integral subunit components of native Kv4 channel complexes that may regulate A-type currents, and hence neuronal excitability, in response to changes in intracellular calcium. The encoded protein also functions as a calcium-regulated transcriptional repressor, and interacts with presenilins. Alternatively spliced transcript variants encoding different isoforms have been described.
Specifications

Specifications

KChIP3
Polyclonal
Unconjugated
KCNIP3
Liquid
KCNIP3
Recombit fragment corresponding to a region within amino acids 1 and 256 of Human CSEN
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Western Blot
0.88 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q9QXT8, Q9JM47, Q9Y2W7
CSEN, DREAM, KCHIP3, A-type potassium channel modulatory protein 3, DRE-antagonist modulator, calsenilin, calsenilin, presenilin-binding protein, EF hand transcription factor, kv channel-interacting protein 3, potassium channel interacting protein 3
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
30818, 65199, 56461
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit