Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GM-CSF, Biotin, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA132896

Catalog No. PIPA132896

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PA1-32896 detects CSF2 from mouse samples.PA1-32896 has been successfully used in Western blot applications.The PA1-32896 immunogen is: Recombinant mouse GM-CSF
Specifications

Specifications

CSF2
Polyclonal
PBS with no preservative
P01587
CSF2
Recombinant mouse GM-CSF
Affinity chromatography
RUO
Antibody
Polyclonal
Murine
ELISA, Western Blot
Biotin
Csf2
Purified
Rabbit
IgG
50μg
Store at -20°C
Primary
12981
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit