Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GM-CSF, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA132122

Catalog No. PA132122

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The protein encoded by this gene is a cytokine that controls the production, differentiation, and function of granulocytes and macrophages. The active form of the protein is found extracellularly as a homodimer. This gene has been localized to a cluster of related genes at chromosome region 5q31, which is known to be associated with interstitial deletions in the 5q- syndrome and acute myelogenous leukemia. Other genes in the cluster include those encoding interleukins 4, 5, and 13.

PA1-32122 detects CSF2 from human samples.PA1-32122 has been successfully used in ELISA and Western blot applications.The PA1-32122 immunogen is: Recombinant human GM-CSF produced in Chinese Hamster cells
Specifications

Specifications

CSF2
Polyclonal
whole serum diluted in 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
P04141
GMCSF, MGC131935, MGC138897
Rabbit
IgG
RUO
Antibody
Polyclonal
Human
ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Radioimmunoassay, Western Blot, Western Blotting
Unconjugated
CSF2
Serum
CSF2
Recombinant human GM-CSF produced in Chinese Hamster cells
500μL
Store at -20°C
Primary
1437
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit