Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CSGALNACT1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524847

Catalog No. PIPA524847


Add to cart

Description

Description

CSGALNACT1 transfers 1,4-N-acetylgalactosamine (GalNAc) from UDP-GalNAc to the non-reducing end of glucuronic acid (GlcUA). CSGALNACT1 required for addition of the first GalNAc to the core tetrasaccharide linker and for elongation of chondroitin chains. CSGALNACT1 is important role in chondroitin chain biosynthesis in cartilage.
Specifications

Specifications

CSGALNACT1
Polyclonal
PBS with 0.09% sodium azide
Q8TDX6, Q8BJQ9
CSGalNAcT-1, ChGn, beta4GalNAcT, beta4GalNAcT-1, chondroitin beta-1,4-N-acetylgalactosaminyltransferase 1, chondroitin beta1,4 N-acetylgalactosaminyltransferase
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
234356, 55790
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
CSGALNACT1
Liquid
CSGALNACT1
KLH conjugated synthetic peptide between 192-220 amino acids from the central region of human CSGALNACT1
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human, Mouse
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit