Learn More
CSK Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA119417
Catalog No. 01-676-754
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
This antibody detects total levels of endogenous Csk protein.
C-terminal Src kinase (Csk) is a non-receptor protein tyrosine kinase. One of its major functions is to specifically phosphorylate a conserved C-terminal tyrosine of Src family kinases thereby down-regulating their kinase activity. CSK is expressed ubiquitously as a protein of approximately 50 KDa in cytosol, but a fraction distributes to lipid rafts due to interaction with the lipid raft-associated protein PAG (Csk-binding protein).
Specifications
Specifications
Csk
Polyclonal
PBS with 15mM sodium azide; pH 7.4
P41240
CSK; c-src tyrosine kinase
Rabbit
IgG
100 μg
4° C, do not freeze
Primary
1445
Western Blot
Unconjugated
CSK
Liquid
CSK
Bacterially expressed recombit protein corresponding to C-terminal part (residues 330-450) of Csk
Protein A
RUO
Antibody
Polyclonal
Human