Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CSPP1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526392

Catalog No. PIPA526392

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

CSPP1 interacts with centrosomes and microtubules and plays a role in cell-cycle progression and spindle organization.
Specifications

Specifications

CSPP1
Polyclonal
PBS with 0.09% sodium azide
Q1MSJ5
CSPP, centrosome and spindle pole-associated protein 1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
79848
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
CSPP1
Liquid
CSPP1
KLH conjugated synthetic peptide between 304-333 amino acids from the N-terminal region of human CSPP1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit