Learn More
cysteine and glycine-rich protein 1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant CSRP1.
Supplier: Abnova Corporation H00001465A01
Description
This gene encodes a member of the cysteine-rich protein (CSRP) family. This gene family includes a group of LIM domain proteins, which may be involved in regulatory processes important for development and cellular differentiation. The LIM/double zinc-finger motif found in this gene product occurs in proteins with critical functions in gene regulation, cell growth, and somatic differentiation. [provided by RefSeq
Sequence: EAPGHRPTTNPNASKFAQKIGGSERCPRCSQAVYAAEKVIGAGKSWHKACFRCAKCGKGLESTTLADKDGEIYCKGCYAKNFGPKGFGFGQGAGALVHSSpecifications
cysteine and glycine-rich protein 1 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant CSRP1. | |
CSRP1 | |
CRP/CRP1/CSRP/CYRP/D1S181E/DKFZp686M148 | |
Mouse | |
50 μL | |
Yes | |
1465 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_004078 | |
CSRP1 | |
CSRP1 (NP_004069, 94 a.a. ∼ 192 a.a) partial recombinant protein with GST tag. | |
RUO | |
Primary | |
Human | |
Serum |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.