Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRP2, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA128122

Catalog No. PIPA128122

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

CSRP2 is a member of the CSRP family of genes, encoding a group of LIM domain proteins, which may be involved in regulatory processes important for development and cellular differentiation. CRP2 contains two copies of the cysteine-rich amino acid sequence motif (LIM) with putative zinc-binding activity, and may be involved in regulating ordered cell growth. Other genes in the family include CSRP1 and CSRP3.

PA1-28122 detects CSRP2 from human, mouse, rat samples.PA1-28122 has been successfully used in Western blot applications.The PA1-28122 immunogen is: Recombinant fragment, corresponding to amino acids 50-125 of Human CSRP2.
Specifications

Specifications

CSRP2
Polyclonal
PBS with no preservative
Q16527, P97314, Q62908
CRP2, LMO5, SmLIM
Chicken
IgY
50μg
Store at -20°C
Primary
1466
Western Blot
Unconjugated
CSRP2
Purified
CSRP2
Recombinant fragment, corresponding to amino acids 50-125 of Human CSRP2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Murine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit