Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cystatin 9 Rabbit anti-Human, Polyclonal, Invitrogen
Rabbit Polyclonal Antibody
Manufacturer: Invitrogen PA524535
Description
May play a role in hematopoietic differentiation or inflammation.Specifications
Cystatin 9 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q5W186 | |
CLM, cystatin-9, cystatin-like molecule | |
Rabbit | |
IgG | |
400 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
128822 |
Flow Cytometry, Immunohistochemistry, Western Blot | |
Unconjugated | |
CST9 | |
Liquid | |
CST9 | |
KLH conjugated synthetic peptide between 131-159 amino acids from the C-terminal region of human CST9 | |
Antigen affinity chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title