Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CT112 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524475

Catalog No. PIPA524475

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with mouse based on sequence homology.

Specifications

Specifications

CT112
Polyclonal
PBS with 0.09% sodium azide
Q96MY1
C20orf113, dJ1184F4.2, dJ1184F4.4, uncharacterized protein C20orf112
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
140688
Flow Cytometry, Western Blot
Unconjugated
NOL4L
Liquid
NOL4L
KLH conjugated synthetic peptide between 210-239 amino acids from the central region of human CT112
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit