Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CT173 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA524315
Description
CT173 Polyclonal antibody specifically detects CT173 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
CT173 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q96LM9 | |
C20orf173 | |
KLH conjugated synthetic peptide between 48-76 amino acids from the central region of human CT173 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
C20orf173 | |
C20orf173; chromosome 20 open reading frame 173; dJ477O4.4; Uncharacterized protein C20orf173 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
140873 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction