Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Actin alpha/gamma Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA132416

Catalog No. PIPA132416


Add to cart

Description

Description

PA1-32416 detects CTB & CTG1 from human, mouse samples. PA1-32416 has been successfully used in Western blot applications. The PA1-32416 immunogen is: The epitope that this antibody recognizes maps to the N-terminus of human beta actin using the numbering given in Swiss-prot entry P60709. The N-terminus of beta actin is highly conserved with gamma actin.

This gene encodes one of six different actin proteins. Actins are highly conserved proteins that are involved in cell motility, structure, and integrity. This actin is a major constituent of the contractile apparatus and one of the two nonmuscle cytoskeletal actins.
Specifications

Specifications

Actin alpha/gamma
Polyclonal
Unconjugated
ACTG1
Liquid
ACTB, ACTG1
The epitope that this antibody recognizes maps to the N-terminus of human beta actin using the numbering given in Swiss-prot entry P60709. The N-terminus of beta actin is highly conserved with gamma actin.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Western Blot
1 mg/mL
tris citrate, tris phosphate with 0.09% sodium azide; pH 7
P60709, P63261, P60710, P63260
PS1TP5BP1, Atcb
Rabbit
IgG
100 μL
4° C, do not freeze
Primary
11461, 11465, 60, 71
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit