Learn More
CtBP1 (C-terminal Binding Protein-1) Mouse, Unlabeled, Clone: 3, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 612042
Description
Transcriptional repression is critical for genetic regulation of many cellular proteins, and involves either a transcription regulator that has an intrinsic repressor domain, or a transcription regulator that contains a binding site in the regulatory domain where a corepressor can bind. C-terminal binding proteins (CtBP1 and CtBP2) are a class of corepressors that interact with a subset of transcription factors through a PLDLSL sequence motif. CtBP1 and CtBP2 are 80% homologous, and both are expressed at high levels in embryonic tissues. However, CtBP1 is expressed higher in adult tissues, and is more widely expressed in embryonic and adult tissues. Both CtBP1 and CtBP2 bind the δEF1 zinc finger-homeodomain transcription factor, and enhance transcriptional repression via interaction with the PLDLSL-sequence in δEF1. CtBP1 and CtBP2 bind other zinc finger transcription factors, including Kheper and BKLF. In addition, CtBP2 interacts with other transcription factors, such as hFOG-2, Evi-1, AREB6, and ZEB. Thus, CtBP1 and CtBP2 may be important corepressors for a variety of transcriptional factors.
Host Species: Mouse
Clone: 3
Isotype: IgG1
Species Reactivity [for Features Main]: Mouse
Immunogen: Mouse CtBP1 aa. 345-441
Immunofluorescence, Western Blotting
Specifications
CtBP1 (C-terminal Binding Protein-1) | |
Monoclonal | |
250μg/mL | |
C-terminal Binding Protein-1 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
Western Blot | |
3 | |
Unconjugated | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Mouse CtBP1 aa. 345-441 | |
50 μg | |
Cell Biology | |
Canine, Chicken, Human, Mouse, Rat | |
IgG1 |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.