Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CtBP1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524409

Catalog No. PIPA524409

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene encodes a protein that binds to the C-terminus of adenovirus E1A proteins. This phosphoprotein is a transcriptional repressor and may play a role during cellular proliferation. This protein and the product of a second closely related gene, CTBP2, can dimerize. Both proteins can also interact with a polycomb group protein complex which participates in regulation of gene expression during development. Alternative splicing of transcripts from this gene results in multiple transcript variants.
Specifications

Specifications

CTBP1
Polyclonal
PBS with 0.09% sodium azide
Q13363, O88712
BARS, C-terminal-binding protein 1, brefeldin A-ribosylated substrate
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
13016, 1487
Western Blot
Unconjugated
CTBP1
Liquid
CTBP1
KLH conjugated synthetic peptide between 409-438 amino acids from the C-terminal region of human CTBP1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit