Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CTCF Mouse anti-Human, Clone: 1.2.1.5.4, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 041506
Description
Anti-CTCF Antibody, clone 1.2.1.5.4 is a Mouse Monoclonal Antibody for detection of CTCF also known as 11-zinc finger protein, CCCTC-binding factor and has been validated in western blot, immunoprecipitation and immunohistochemistry.Specifications
CTCF | |
Monoclonal | |
Unconjugated | |
NP_006556.1 | |
Mouse | |
Protein G purfied | |
Primary | |
2-8°C for one year | |
IgG1 κ |
Immunohistochemistry, Immunoprecipitation, Western Blot | |
1.2.1.5.4 | |
Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide. | |
CTCF | |
His-tagged recombinant protein corresponding to human CTCF. | |
100 μg | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only