Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTCF Mouse, Unlabeled, Clone: 48, BD

Mouse Monoclonal Antibody

$232.00 - $427.00

Specifications

Antigen CTCF
Isotype IgG1
Immunogen Human CTCF aa. 184-290
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB612148
View Documents
BD Biosciences
612148
50μg
Each for $232.00
Add to cart
 
BDB612149
View Documents
BD Biosciences
612149
150μg
Each for $427.00
Add to cart
 
Description

Description

The family of nucleic acid-binding C2H2 type zinc finger transcription factors is divided into two classes. One class consists of small proteins (Gli1, Krox-20, WT1, Egr-1, and Sp1) with conserved zinc finger clusters of 3 to 5 units, while the other class (ZNF91, ZNF74, ZFP37, CTCF) can contain more than 10 zinc finger clusters. CTCF is a ubiquitously expressed, highly conserved transcription factor that contains 10 C2H2- and 1 C2H-type zinc-finger motifs. CTCF binds to and represses transcription at the promoter-proximal regions of c-myc oncogenes,
while CTCF can bind to and activate transcription at the promoter for amyloid β. Another mechanism of CTCF-mediated repression may include binding to insulator regions between enhancers and promoters resulting in enhancer blocking. In addition, CTCF gene may be a candidate tumor suppressor gene, since it localizes to a narrow cancer-associated chromosome region, and has been shown to have tumor-specific rearrangements in breast cancer patients. Thus, CTCF transcriptional regulation may be important for cell cycle progression, differentiation, apoptosis, and tumorigenesis.

Immunofluorescence, Western Blotting

Specifications

Specifications

CTCF
Human CTCF aa. 184-290
Unconjugated
Cell Biology
Mouse
Affinity Purified
Monoclonal
Store undiluted at -20°C.
IgG1
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Primary
Western Blot
RUO
48
250μg/mL
SDS
Product Certifications

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit