Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTCF Rabbit anti-Human, Mouse, Bovine, Chicken, Equine, Rabbit, Zebrafish, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP178106

Catalog No. NBP178106

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

CTCF Polyclonal antibody specifically detects CTCF in Human, Mouse, Bovine, Chicken, Equine, Rabbit, Zebrafish samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications

Specifications

CTCF
1 mg/ml
Western Blot 1:500-1:2:000, ELISA 1:10000-1:50000, Immunohistochemistry 5 ug/mL
P49711
11 zinc finger transcriptional repressor, 11-zinc finger protein, CCCTC-binding factor, CCCTC-binding factor (zinc finger protein), CTCFL paralog, transcriptional repressor CTCF
Rabbit
IgG
0.1 mg
Breast Cancer, Cancer, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Immune System Diseases, Immunology, Transcription Factors and Regulators
Primary
10664
Bovine, Chicken, Equine, Human, Mouse, Rabbit, Zebrafish
ELISA, Immunohistochemistry, Western Blot
Unconjugated
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Affinity Purified
CTCF
CTCF affinity purified antibody was prepared by repeated immunizations with a synthetic peptide corresponding to a region near the C-terminus of CTCF protein.
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
This antibody reacts with endogenous CTCF protein. A BLAST analysis was used to suggest reactivity with CTCF from human, mouse, horse, bovine, panda, rabbit.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only