Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTDSPL Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523546

Catalog No. PIPA523546


Add to cart

Description

Description

The synaptonemal complex is a proteinaceous complex that apparently mediates synapses during the zygotene stage and then disintegrates. It is a complex structure that unites homologous chromosomes during the prophase stage of meiosis. It is the result of
Specifications

Specifications

CTDSPL
Polyclonal
PBS with 0.09% sodium azide
O15194
C3orf8, HYA22, PSR1, RBSP3, SCP3, CTD small phosphatase-like protein, CTDSP-like, NIF-like protein, NLI-interacting factor 1, RB protein serine phosphatase from chromosome 3, carboxy-terminal domain RNA polymerase II polypeptide A small phosphatase 3, nuclear LIM interactor-interacting factor 1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10217
Flow Cytometry, Western Blot
Unconjugated
CTDSPL
Liquid
CTDSPL
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human CTDSPL
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit