Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTGF Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA122376

Catalog No. PA122376


Add to cart

Description

Description

PA1-22376 detects CTGF in Human, Mouse and Rat samples. PA1-22376 has been successfully used in Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) and Western Blot procedures. PA1-22376 immunogen corresponds to Recombit fragment, corresponding to amino acids 223-348 of Mouse CTGF. Store at 4°C short term. For extended storage aliquot and store at -20°C or below. Avoid freeze/thaw cycles.

CTGF (connective tissue growth factor) is a 38 kD, cysteine-rich, secreted peptide. It is a new member of the peptide family that include serum-induced immediate early gene products, a v-src-induced peptide, and a putative proto-oncogene. Among the many functions of the CTGF gene family are embryogenesis, wound healing, and regulation of extracellular matrix production.
Specifications

Specifications

CTGF
Polyclonal
Unconjugated
CTGF
Liquid
CTGF
Recombit fragment, corresponding to amino acids 223-348 of Mouse CTGF.
Protein A
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 0.1% sodium azide
P29279, P29268, Q9R1E9
CCN2, HCS24, IGFBP8, MGC102839, NOV2
Rabbit
IgG
50 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
14219, 1490, 64032
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit