Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

CTLA-4 Mouse anti-Human, Clone: 48815, R&D Systems™

Mouse Monoclonal Antibody

Specifications

Antigen CTLA-4
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human CTLA-4 Ala37-Phe162 (Thr174Ala) Accession # Q6GR94
Clone 48815
Applications Western Blot
Conjugate Unconjugated
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB325SP
View Documents
R&D Systems
MAB325SP
25 ug
Each for $125.00
N/A
MAB325100
View Documents
R&D Systems
MAB325100
100 ug
Each for $377.00
N/A
MAB325500
View Documents
R&D Systems
MAB325500
500 ug
Each for $839.00
N/A
Description

Description

CTLA-4 Monoclonal antibody MAB325 Specifically detects CTLA-4 in Human samples. It is validated for Western Blotting
Specifications

Specifications

CTLA-4
48815
Unconjugated
Q6GR94
Monoclonal
Primary
Detects human CTLA-4 in direct ELISAs and Western blots. Shows approximately 10% cross-reactivity with recombinant mouse CTLA-4 and no cross-reactivity with recombinant human (rh) CD28 or rhICOS.
Protein A or G purified from hybridoma culture supernatant
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human CTLA-4 Ala37-Phe162 (Thr174Ala) Accession # Q6GR94
Western Blot
Purified
Mouse
IgG2b
1493
RUO
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit