Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha Catenin Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA513526

Catalog No. PIPA513526

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine and rabbit based on sequence homology.

Alpha catenin is a member of the b-catenin family of cytosolic proteins. Alpha catenin interacts with N-cadherin and b-catenin on the cytosolic side of adherins junctions to mediate the interaction with the actin cytoskeleton. Alpha catenin is a homodimer in solution, but dissociates to form a 1:1 heterodimer with b-catenin in order to interact with cadherins. It also regulates morphological plasticity of synapses and cerebellar and hippocampal lamination during development. Alpha catenin also functions in the control of the startle modulation.
Specifications

Specifications

alpha Catenin
Polyclonal
PBS with 0.09% sodium azide
P35221, P26231
Catenin alpha-1; Cadherin-associated protein; Alpha E-catenin; NY-REN-13 antigen; CTNNA1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
12385, 1495
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
CTNNA1
Liquid
CTNNA1
KLH conjugated synthetic peptide between 30-59 amino acids from the N-terminal region of human CTNA1
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit