Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha Catenin, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA127905

Catalog No. PIPA127905

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Associates with the cytoplasmic domain of a variety of cadherins. The association of catenins to cadherins produces a complex which is linked to the actin filament network. and which seems to be of primary importance for cadherins cell-adhesion properties

PA1-27905 detects CTNNA1 from human, mouse, rat, and chimpanzee samples.PA1-27905 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The PA1-27598 immunogen is a synthetic peptide corresponding to human Alpha-1 catenin.
Specifications

Specifications

CTNNA1
Polyclonal
PBS with no preservative
P35221, P26231
CAP102, FLJ36832, FLJ52416
Chicken
IgY
50μg
Store at -20°C
Primary
1495
Western Blot
Unconjugated
CTNNA1
Purified
CTNNA1
Synthetic peptide corresponding to human Alpha-1 catenin.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Murine, Non-human Primate, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit