Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

catenin (cadherin-associated protein), delta 1, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human CTNND1 protein.

Manufacturer:  Abnova Corporation H00001500B01

Catalog No. 89-005-197


Add to cart

Description

Description

This gene encodes a member of the Armadillo protein family, which function in adhesion between cells and signal transduction. Multiple translation initiation codons and althernative splicing result in many different isoforms being translated. Not all of the full-length natures of the described transcript variants have been determined. [provided by RefSeq

Sequence: MIGEEVPSDQYYWAPLAQHERGSLASLDSLRKGGPPPPNWRQPELPEVIAMLGFRLDAVKSNAAAYLQHLCYRNDKVKTDVRKLKGIPVLVGLLDHPKKEVHLGACGALKNISFGRDQDNKIAIKNCDGVPALVRLLRKARDMDLTEVITGTLWNLSSHDSIKMEIVDHALHALTDEVIIPHSGWEREPNEDCKPRHIEWESVLTNTAGCLRNVSSERSEARRKLRECDGLVDALIFIVQAEIGQKDSDSKLVENCVCLLRNLSYQVHREIPQAERYQEAAPNVANNTGPHAASCFGAKKGKGKKPIEDPANDTVDFPKRTSPARGYELLFQPEVVRIYISLLKESKTPAILEASAGAIQNLCAGRWTYGRYIRSALRQEKALSAIADLLTNEHERVVKAASGALRNLAVDARNKELIGKHAIPNLVKNLPGGQQNSSWNFSEDTVISILNTINEVIAENLEAAKKLRETQGIEKLVLINKSGNRSEKEVRAAALVLQTIWGYKELRKPLEKEGWKKSDFQVNLNNASRSQSSHSYDDSTLPLIDRNQKSDKKPDREEIQMSNMGSNTKSLDNNYSTPNERGDHNKTLDRSGDLGDMEPLKGTTPLMQKI
Specifications

Specifications

catenin (cadherin-associated protein), delta 1
Unconjugated
No additive
BC010501
CAS|CTNND|KIAA0384|P120CAS|P120CTN|p120
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
1500
Immunofluorescence, Western Blot
Mouse polyclonal antibody raised against a full-length human CTNND1 protein.
CTNND1
Serum
CTNND1
CTNND1 (AAH10501, 1 a.a. ~ 610 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit