Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cathepsin D Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526457

Catalog No. PIPA526457

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

CTSD encodes a lysosomal aspartyl protease composed of a dimer of disulfide-linked heavy and light chains, both produced from a single protein precursor. This proteinase, which is a member of the peptidase C1 family, has a specificity similar to but narrower than that of pepsin A. Transcription of this gene is initiated from several sites, including one which is a start site for an estrogen-regulated transcript.
Specifications

Specifications

Cathepsin D
Polyclonal
PBS with 0.09% sodium azide
P07339
CLN10, CPSD, ceroid-lipofuscinosis, neuronal 10, lysosomal aspartyl peptidase, lysosomal aspartyl protease
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
1509
Western Blot
Unconjugated
CTSD
Liquid
CTSD
KLH conjugated synthetic peptide between 377-408 amino acids from the C-terminal region of human CTSD
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit