Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTTNBP2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525404

Catalog No. PIPA525404

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

CTTB2 is a protein with six ankyrin repeats andseveral proline-rich regions. A similar gene in rat interacts witha central regulator of the actin cytoskeleton.
Specifications

Specifications

CTTNBP2
Polyclonal
PBS with 0.09% sodium azide
Q8WZ74
C7orf8, CORTBP2, Orf4, cortactin-binding protein 2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
83992
Immunohistochemistry, Western Blot
Unconjugated
CTTNBP2
Liquid
CTTNBP2
KLH conjugated synthetic peptide between 72-100 amino acids from the N-terminal region of human CTTB2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit