Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C21orf2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524270

Catalog No. PIPA524270

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

C21orf2
Polyclonal
PBS with 0.09% sodium azide
O43822
A2, YF5, YF5/A2, nuclear encoded mitochondrial protein, cDNA A2-YF5, uncharacterized protein C21orf2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
755
Flow Cytometry, Western Blot
Unconjugated
C21orf2
Liquid
C21ORF2
KLH conjugated synthetic peptide between 169-198 amino acids from the C-terminal region of human CU002
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit