Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPATC1L Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525135

Catalog No. PIPA525135


Add to cart

Description

Description

The function of this protein has not been specifically defined.
Specifications

Specifications

SPATC1L
Polyclonal
PBS with 0.09% sodium azide
Q9H0A9, Q9D9W0
uncharacterized protein C21orf56
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
76573, 84221
Western Blot
Unconjugated
SPATC1L
Liquid
SPATC1L
KLH conjugated synthetic peptide between 226-254 amino acids from the C-terminal region of human CU056
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit