Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CUEDC2, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP177334

Catalog No. NBP177334

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Specifications

Specifications

CUEDC2
1 mg/mL
Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunofluorescence
NP_076945
bA18I14.5, C10orf66, chromosome 10 open reading frame 66, CUE domain containing 2, CUE domain-containing protein 2, MGC2491
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
79004
Human, Mouse, Rat
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
PBS with 0.02% Sodium Azide
Affinity Purified
CUEDC2
Antibody was raised against a 19 amino acid synthetic peptide from near the carboxy terminus of human CUEDC2. The immunogen is located within the last 50 amino acids of CUEDC2.
34 kDa
0.1 mg
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
CUEDC2 antibody will not cross-react with CUEDC1.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only