Learn More
CUGBP1 Mouse anti-Bovine, Human, Mouse, Pig, Rat, Rabbit, Clone: 3B1, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05621
Catalog No. 05-621-MI
Add to cart
Description
Description
Anti-CUGBP1 Antibody, clone 3B1 is a high quality Mouse Monoclonal Antibody for the detection of CUGBP1 and has been validated in EMSA, immunoprecipitation, western blot, immunocytochemistry.
Specifications
Specifications
CUGBP1
3B1
0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Purified
Murine
IgG
200μg
Primary
Bovine, Human, Murine, Porcine, Rabbit, Rat
Electromobility Shift Assay, Immunocytochemistry, Immunoprecipitation, Western Blot
Unconjugated
NM_006560 and others
CUGBP1, BRUNOL2, CELF-1, CUGBP, NAB50, CELF1, CUG-BP, hNab50, CUG-BP1
Full-length GST fusion protein corresponding to human CUG-BP1, also known as heterogeneous nuclear ribonucleoprotein (hnRNP) hNab50
Protein G chromatography
−20°C for 24 months.
Monoclonal