Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CUGBP2 Mouse anti-Human, Clone: 1H2, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 04047
Description
This Anti-CUGBP2 Antibody, clone 1H2 is validated for use in western blotting for the detection of CUGBP2.Specifications
CUGBP2 | |
Monoclonal | |
Unconjugated | |
NM_001025076.2; NM_001025077.2; NM_001083591.1; NM_006561.3 | |
Mouse | |
Protein A purfied | |
Primary | |
−20°C for 24 months. | |
IgG1 |
Western Blot | |
1H2 | |
100μg of Protein A purified mouse monoclonal IgG1 in 100μL of 1X PBS, pH 7.0, 0.1% Azide. | |
CUGBP2, CUG-BP2, BRUNOL3, CELF2, CELF-2, Etr-3, ETR-3, ETR3, NAPOR-2, NAPOR2, NAPOR, hNAPOR | |
A synthetic peptide corresponding to amino acids 209-415 of human CuGBP2. | |
100 μg | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only