Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CUL-1 Mouse, Unlabeled, Clone: 19, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 612040

Catalog No. BDB612040


Add to cart

Description

Description

The cullins are a family of proteins that are integral to cell cycle regulation. Members of this family include human Cul proteins, C.elegans Ce-Cul-1, and S. cerevisiae Cdc53. Proper control of cell cycle progression is essential for the prevention of tumorigenesis. Mutant forms of both Ce-Cul-1 and Cdc53 have been implicated in the oncogenic process. The human Cul family of proteins, Cul-1, -2, -3, -4A, -4B, and -5, have also been implicated in oncogenic processes. Abnormal nuclear localization of Cul-2 is seen in the VHL (von Hippel-Lindau) syndrome, which includes development of a variety of cancers. Cul-3 is widely expressed in normal cells, and has increased expression in colon cancer cells. Cul-3 can associate with cyclin E in mammalian cells, and overexpression of Cul-3 targets cyclin E for ubiquination. Cul-1 associates with the ubiquination-conjugating complex, cyclin A/CDC34/p45[SKP2]/p19[SKP1]. In addition, Cul-1 interacts with p19[SKP1], E2 ubiquination-conjugating enzyme, and FWD1 to facilitate ubiquitin degradation of IκBα, β.catenin, and Vpu proteins. Thus, Cul-1, as well as other cullin family members, may have important roles in cycle-dependent ubiquitin degradation.

Host Species: Mouse
Clone: 19
Isotype: IgG1
Species Reactivity [for Features Main]: Mouse
Immunogen: Mouse CUL-1 aa. 56-246

Immunofluorescence, Western Blotting

Specifications

Specifications

CUL-1
19
Unconjugated
Affinity Purified
Mouse CUL-1 aa. 56-246
50μg
Cell Biology
Primary
Canine, Chicken, Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit