Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CUL-2 Mouse, Unlabeled, Clone: 4, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610778

Catalog No. BDB610778


Add to cart

Description

Description

The cullins are a family of proteins that are integral to cell cycle regulation. Members of this family include human Cul-2, C.elegans Ce-Cul-1, and S. cerevisiae Cdc53. Proper control of cell cycle progression is essential for the prevention of tumorigenesis. Mutant forms of both Ce-Cul-1 and Cdc53 have been implicated in the oncogenic process. Abnormal nuclear localization of Cul-2 is seen in a rare hereditary condition known as VHL (von Hippel-Lindau) syndrome. Patients with VHL syndrome possess a mutant VHL gene and develop cancers such as retinal anginomas, CNS hemangioblastomas, and renal cell carcinomas. In normal cells, the VHL protein competes with subunit A of elongin, a transcription elongation factor, for binding to subunits B and C. The formation of the VHL-B-C complex inhibits elongation. Additional data indicates that this complex associates with CUL-2 and promotes CUL-2 nuclear translocation. In cells with mutated VHL, transcriptional elongation is not efficiently curtailed and CUL-2 is not properly localized in the nucleus. Therefore, CUL-2 mediates VHL function, suggesting CUL-2 may be a candidate human tumor suppressor.

Immunofluorescence, Immunoprecipitation, Western Blotting

Specifications

Specifications

CUL-2
4
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Western Blot
250μg/mL
Cullin-2
Affinity Purified
Human CUL-2 aa. 18-182
50μg
Cell Biology
Primary
Canine, Human, Murine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit