Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cullin 1, Polyclonal, Invitrogen™

Catalog No. PIPA124260
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
PIPA124260 100 μg
1 options

Catalog No. PIPA124260

Supplier: Thermo Scientific PA124260

Rabbit Polyclonal Antibody

Cullin 1 is a member of the family of human cullin genes (CUL 1, 2, 3, 4a, 4b, and 5) homologous to the S. cerevisiae cdc 53 gene. CUL 1 forms a complex with human p19 skp1 and F box protein p45 skp2, both of which, together with cdc 53 (known as CUL A), as three subunits form a ubiquitin ligase controlling proteolysis of G1 cell cycle regulatory proteins. The CUL 1/p19 skp1/p45 skp2complex is thought to play a role in the ubiquitination of G1 regulatory proteins.

PA1-24260 detects Cullin 1 (CUL-1) in human samples.PA1-24260 has been successfully used in Western blot procedures.The PA1-24260 immunogen is a synthetic peptide corresponding to residues HQQLLGEVLTQLSSRFKPRVPVIKKCIDILIEKEYLERVDGEKDTYSYLA of human CUL1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cullin 1
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Gene CUL1
Gene Accession No. Q13616
Gene Alias CUL-1
Gene Symbols CUL1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues HQQLLGEVLTQLSSRFKPRVPVIKKCIDILIEKEYLERVDGEKDTYSYLA of human CUL1.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8454
Target Species Human
Content And Storage Store at -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.