Learn More
Cullin 3, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA129163
Catalog No. PIPA129163
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Cullin 3 is a member of the family of human cullin genes (CUL 1, 2, 3, 4a, 4b, and 5) homologous to the S. cerevisiae cdc 53 gene. CUL 1 forms a complex with human p19 skp1 and F box protein p45 skp2. Together with cdc 53 (Known as Cul A), as three subuni
PA1-29163 detects Cullin 3 from mouse, human samples.PA1-29163 has been successfully used in ICC, IF and IP applications.The PA1-29163 immunogen is: Synthetic peptide corresponding to the N-terminus of human Cullin-3 protein.
Specifications
Specifications
Cullin 3
Polyclonal
Unconjugated
CUL3
Liquid
CUL3
Synthetic peptide corresponding to the N-terminus of human Cullin-3 protein.
Protein A
RUO
Antibody
Polyclonal
Human, Murine
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
1mg/mL
PBS with 0.2% BSA and 0.1% sodium azide; pH 7.4
Q13618, Q9JLV5
CUL-3
Rabbit
IgG
500μL
4° C, do not freeze
Primary
26554, 8452