Learn More
anti-CX3CL1/Fractalkine, Alexa Fluor 488, Clone: 126315, R&D Systems™
Rat Monoclonal Antibody has been used in 1 publication
Manufacturer:  R&D Systems FAB571G025
Catalog No. FAB571G025
This item is not returnable. View return policy
Description
Description
CX3CL1/Fractalkine Monoclonal specifically detects CX3CL1/Fractalkine in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
CX3CL1/Fractalkine
126315
Flow Cytometry 5 uL/10^6 cells
Supernatant
CX3CL1
Mouse myeloma cell line NS0-derived recombinant mouse CX3CL1/Fractalkine, S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse CX3CL1/Fractalkine, and E. coli-derived recombinant mouse CX3CL1/Fractalkine
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
6376
Mouse
Flow Cytometry
Alexa Fluor 488
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
ABCD-3, C3Xkine, chemokine (C-X3-C motif) ligand 1, CX3C membrane-anchored chemokine, CXC3, CXC3C, FKN, fractalkine, Neurotactin, neurotactin), NTNsmall inducible cytokine subfamily D (Cys-X3-Cys), member 1 (fractalkine, NTTSmall-inducible cytokine D1, SCYD1C-X3-C motif chemokine 1, small inducible cytokine subfamily D (Cys-X3-Cys), member-1
Rat
IgG2a
25 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects mouse CX3CL1/Fractalkine in Western blots. In direct ELISAs, this antibody shows 100% cross-reactivity with the chemokine domain of recombinant human (rh) CX3CL1, 13% cross-reactivity with rhCX3CL1 (full length), and 50% cross-reactivity with recombinant rat CX3CL1.